Gene Transcripts

Gene Symbol
Ccm2
Entrez Gene ID
216527
Gene Name
cerebral cavernous malformation 2
Gene Aliases
BC029157
Chromosome Location
Chr. 11: 6546887 - 6596761 on Build GRCm38
UniGene ID
Mm.221271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190343.1 NP_001177272.1 8, 9 1023
NM_001190344.1 NP_001177273.1 9, 10 1109
NM_146014.3 NP_666126.1 9, 10 1197
GenBank mRNA AK188818.1 - 1, 2 22
BC029157.1 - 9, 10 1128 4952288

View Full Product Details