Gene Transcripts

Gene Symbol
Slc17a2
Entrez Gene ID
218103
Gene Name
solute carrier family 17 (sodium phosphate), member 2
Gene Aliases
C730032N17Rik, NPT3
Chromosome Location
Chr. 13: 23806941 - 23825191 on Build GRCm38
UniGene ID
Mm.24030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144836.2 NP_659085.1 3 625
GenBank mRNA AK040717.1 - 3 626
AK050184.1 - 3 608
AK050277.1 - 4 589
BC018306.1 - 2 263 4193755

View Full Product Details