Gene Transcripts

Gene Symbol
Rfesd
Entrez Gene ID
218341
Gene Name
Rieske (Fe-S) domain containing
Gene Aliases
AI256775, D030068K09
Chromosome Location
Chr. 13: 76001535 - 76018712 on Build GRCm38
UniGene ID
Mm.266515
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131068.1 NP_001124540.1 4 878
NM_001131069.1 NP_001124541.1 4 341
NM_178916.5 NP_849247.3 4 1328
GenBank mRNA AK029602.1 - 4 1154
AK040255.1 - 4 833
AK083705.1 - 4 1329
BC030381.1 - 4 953 5375753
CD769672.1 - 4 345
CN839985.1 - 4 351

View Full Product Details