Gene Transcripts
- Gene Symbol
- Tspan7
- Entrez Gene ID
- 21912
- Gene Name
- tetraspanin 7
- Gene Aliases
- 1200014P11Rik, A15, AI323365, Cd231, Mxs1, PE31, R74651, TALLA, Tm4sf2
- Chromosome Location
- Chr. X: 10485116 - 10596604 on Build GRCm38
- UniGene ID
- Mm.18590
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019634.2 |
NP_062608.2
|
2, 3 |
306 |
|
GenBank mRNA |
AF052492.1 |
- |
2, 3 |
254 |
|
AK004776.1 |
- |
2, 3 |
266 |
|
AK147306.1 |
- |
2, 3 |
268 |
|
AK147404.1 |
- |
2, 3 |
270 |
|
AK214746.1 |
- |
1, 2 |
71 |
|
BC026516.1 |
- |
1, 2 |
105 |
4911317 |
BC050153.1 |
- |
2, 3 |
254 |
|
BC058603.1 |
- |
2, 3 |
262 |
6529011 |
BY238972.1 |
- |
2, 3 |
306 |
|
D26483.1 |
- |
2, 3 |
254 |
|
View Full Product Details