Gene Transcripts

Gene Symbol
Txn1
Entrez Gene ID
22166
Gene Name
thioredoxin 1
Gene Aliases
ADF, AW550880, Trx1, Txn
Chromosome Location
Chr. 4: 57943373 - 57956411 on Build GRCm38
UniGene ID
Mm.260618
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011660.3 NP_035790.1 2 267
GenBank mRNA AK007537.1 - 2 148
AK007790.1 - 2 145
BC010756.1 - 2 122 4216106
BC094415.1 - 2 129 3376881
X77585.1 - 2 140

View Full Product Details