Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009507.3 | NP_033533.1 | 3 | 581 | |
GenBank mRNA | AK159238.1 | - | 3 | 602 | |
AK161408.1 | - | 3 | 578 | ||
AK171264.1 | - | 3 | 583 | ||
AK171604.1 | - | 3 | 591 | ||
BC052417.1 | - | 3 | 580 | 6402536 | |
S76748.1 | - | 3 | 581 | ||
U12570.1 | - | 3 | 597 |