Gene Transcripts

Gene Symbol
Prr22
Entrez Gene ID
100504446
Gene Name
proline rich 22
Gene Aliases
Gm546
Chromosome Location
Chr. 17: 56770276 - 56772134 on Build GRCm38
UniGene ID
Mm.46157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195673.1 NP_001182602.1 3 538
GenBank mRNA BC172076.1 - 1 1073

View Full Product Details