Gene Transcripts
- Gene Symbol
- Srsf7
- Entrez Gene ID
- 225027
- Gene Name
- serine/arginine-rich splicing factor 7
- Gene Aliases
- 35kDa, 9430065L19Rik, 9G8, NX-96, Sfrs7
- Chromosome Location
- Chr. 17: 80200080 - 80207318 on Build GRCm38
- UniGene ID
- Mm.292016
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195485.1 |
NP_001182414.1
|
4, 5 |
560 |
|
NM_001195486.1 |
NP_001182415.1
|
4, 5 |
560 |
|
NM_001195487.1 |
NP_001182416.1
|
4, 5 |
560 |
|
NM_146083.2 |
NP_666195.1
|
4, 5 |
560 |
|
GenBank mRNA |
AI550251.1 |
- |
2, 3 |
138 |
|
AK034952.1 |
- |
5, 6 |
1022 |
|
AK045884.1 |
- |
4, 5 |
722 |
|
AK082848.1 |
- |
4, 5 |
560 |
|
AK137795.1 |
- |
4, 5 |
561 |
|
AK161652.1 |
- |
4, 5 |
559 |
|
AK168355.1 |
- |
4, 5 |
560 |
|
AK185991.1 |
- |
1, 2 |
19 |
|
BC014857.1 |
- |
4, 5 |
541 |
2646366 |
BC025529.1 |
- |
4, 5 |
551 |
5342587 |
BC027391.1 |
- |
3, 4 |
1422 |
4945988 |
BY740981.1 |
- |
4, 5 |
559 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK134283.1 |
- |
|
207 |
|
Chr. - - [-] |
View Full Product Details