Gene Transcripts

Gene Symbol
Nkx2-4
Entrez Gene ID
228731
Gene Name
NK2 homeobox 4
Gene Aliases
1700001P03Rik, Nkx-2.4, tinman
Chromosome Location
Chr. 2: 147083337 - 147085439 on Build GRCm38
UniGene ID
Mm.315498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023504.1 NP_075993.1 2 621
GenBank mRNA AF202039.1 - 2 418
AK005620.1 - 1 251

View Full Product Details