Gene Transcripts

Gene Symbol
Flg2
Entrez Gene ID
229574
Gene Name
filaggrin family member 2
Gene Aliases
EG229574
Chromosome Location
Chr. 3: 93197273 - 93221377 on Build GRCm38
UniGene ID
Mm.10755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013804.1 NP_001013826.1 1668

View Full Product Details