Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008725.2 | NP_032751.1 | 2 | 512 | |
GenBank mRNA | AK147180.1 | - | 2 | 516 | |
BC089615.1 | - | 2 | 512 | 6755676 | |
KC526925.1 | - | 1 | 570 | ||
KC526926.1 | - | 1 | 570 | ||
KC526927.1 | - | 1 | 570 |