Gene Transcripts
- Gene Symbol
- Pdap1
- Entrez Gene ID
- 231887
- Gene Name
- PDGFA associated protein 1
- Gene Aliases
- HASPP28, PAP, PAP1
- Chromosome Location
- Chr. 5: 145128770 - 145140089 on Build GRCm38
- UniGene ID
- Mm.188851
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033313.3 |
NP_001028485.1
|
4, 5 |
446 |
|
GenBank mRNA |
AK147170.1 |
- |
4, 5 |
447 |
|
AK161518.1 |
- |
3, 4 |
575 |
|
AK169233.1 |
- |
4, 5 |
380 |
|
BC007162.1 |
- |
3, 4 |
346 |
|
BC055927.1 |
- |
4, 5 |
360 |
|
BC106168.1 |
- |
3, 4 |
334 |
|
BC114598.1 |
- |
2, 3 |
180 |
|
BC139204.1 |
- |
4, 5 |
399 |
8862226 |
BC139205.1 |
- |
4, 5 |
399 |
8862227 |
View Full Product Details