Gene Transcripts

Gene Symbol
Adamts17
Entrez Gene ID
233332
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 17
Gene Aliases
AU023434
Chromosome Location
Chr. 7: 66839723 - 67153171 on Build GRCm38
UniGene ID
Mm.445763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033877.4 NP_001029049.2 3 623
GenBank mRNA AK045148.1 - 3 624
AK053131.1 - 3 624
BC141170.1 - 3 575

View Full Product Details