Gene Transcripts

Gene Symbol
Crebzf
Entrez Gene ID
233490
Gene Name
CREB/ATF bZIP transcription factor
Gene Aliases
1110034C16Rik, 6330417B10Rik, LAZip, LAZipII, SMILE, Zf
Chromosome Location
Chr. 7: 90442735 - 90448043 on Build GRCm38
UniGene ID
Mm.29496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145151.3 NP_660133.2 1 752
GenBank mRNA AF408398.1 - 1 424

View Full Product Details