Gene Transcripts

Gene Symbol
Asphd1
Entrez Gene ID
233879
Gene Name
aspartate beta-hydroxylase domain containing 1
Gene Aliases
A830007L07Rik, Gm168
Chromosome Location
Chr. 7: 126945981 - 126949886 on Build GRCm38
UniGene ID
Mm.444143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039645.1 NP_001034734.1 4 394
GenBank mRNA AK043553.1 - 4 589
AK134834.1 - 3 1085
BC111101.1 - 4 394 40049679

View Full Product Details