Gene Transcripts

Gene Symbol
Dmxl1
Entrez Gene ID
240283
Gene Name
Dmx-like 1
Gene Aliases
C630007L23Rik, D030030C22
Chromosome Location
Chr. 18: 49832993 - 49965473 on Build GRCm38
UniGene ID
Mm.257790
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081371.2 NP_001074840.2 28 7059
GenBank mRNA AK049893.1 - 5 698
AY590892.1 - 28 7059
BC056490.1 - 5 1223

View Full Product Details