Gene Transcripts

Gene Symbol
Ccdc85b
Entrez Gene ID
240514
Gene Name
coiled-coil domain containing 85B
Gene Aliases
AI842788, BB115872
Chromosome Location
Chr. 19: 5453163 - 5457563 on Build GRCm38
UniGene ID
Mm.329657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198616.4 NP_941018.1 1 313
GenBank mRNA AK140354.1 - 1 645
BC058411.1 - 1 299 5710206
BQ445696.1 - 1 299
BY124251.1 - 1 313

View Full Product Details