Gene Transcripts
- Gene Symbol
- Tnfsf13b
- Entrez Gene ID
- 24099
- Gene Name
- tumor necrosis factor (ligand) superfamily, member 13b
- Gene Aliases
- BAFF, BLyS, D8Ertd387e, TALL-1, TALL1, THANK, TNFSF20, zTNF4
- Chromosome Location
- Chr. 8: 10006192 - 10119693 on Build GRCm38
- UniGene ID
- Mm.28835
- Species
- Mus musculus
- Gene Symbol
- Gm10703
- Entrez Gene ID
- 100038550
- Gene Name
- predicted gene 10703
- Gene Aliases
- ENSMUSG00000074477
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.28835
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141839.1 |
- |
1 |
590 |
|
View Full Product Details