Gene Transcripts

Gene Symbol
Lsm14b
Entrez Gene ID
241846
Gene Name
LSM14 homolog B (SCD6, S. cerevisiae)
Gene Aliases
9430078B04, BC040823
Chromosome Location
Chr. 2: 180024902 - 180035466 on Build GRCm38
UniGene ID
Mm.21577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177727.4 NP_808395.2 7, 8 1193
GenBank mRNA AK035040.1 - 7, 8 1222
AK162234.1 - 2, 3 310
BC040823.1 - 7, 8 1193
BC099395.1 - 4, 5 612 30933544
BY720115.1 - 2, 3 308
DV654874.1 - 1, 2 142

View Full Product Details