Gene Transcripts

Gene Symbol
Frem2
Entrez Gene ID
242022
Gene Name
Fras1 related extracellular matrix protein 2
Gene Aliases
6030440P17Rik, 8430406N05Rik, Gm409, b2b1562Clo, my, ne, nv1
Chromosome Location
Chr. 3: 53513938 - 53657912 on Build GRCm38
UniGene ID
Mm.38378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172862.3 NP_766450.2 1 4865
GenBank mRNA AB236662.1 - 1 4865
AJ833643.1 - 1 4594

View Full Product Details