Gene Transcripts

Gene Symbol
Leng9
Entrez Gene ID
243813
Gene Name
leukocyte receptor cluster (LRC) member 9
Gene Aliases
9530024C23Rik, F630035L11Rik
Chromosome Location
Chr. 7: 4148183 - 4149872 on Build GRCm38
UniGene ID
Mm.347643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175529.3 NP_780738.1 1 466
GenBank mRNA AK089196.1 - 1 466
AK090365.1 - 1 83
AK134559.1 - 1 87
AK143899.1 - 78
AK147019.1 - 968
AK164490.1 - 1 425
BC116676.1 - 1 361 40106558

View Full Product Details