Gene Transcripts

Gene Symbol
Shank1
Entrez Gene ID
243961
Gene Name
SH3/ankyrin domain gene 1
Gene Aliases
Chromosome Location
Chr. 7: 44307848 - 44358353 on Build GRCm38
UniGene ID
Mm.360368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034115.1 NP_001029287.1 24 5492
GenBank mRNA AK082624.1 - 2 2872

View Full Product Details