Gene Transcripts

Gene Symbol
Mctp2
Entrez Gene ID
244049
Gene Name
multiple C2 domains, transmembrane 2
Gene Aliases
Gm489
Chromosome Location
Chr. 7: 72077827 - 72306595 on Build GRCm38
UniGene ID
Mm.217412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024703.1 NP_001019874.1 22 2713
GenBank mRNA BC086658.1 - 22 2713

View Full Product Details