Gene Transcripts

Gene Symbol
Spag4
Entrez Gene ID
245865
Gene Name
sperm associated antigen 4
Gene Aliases
1700041K21Rik, MNCb-0953, Sun4, mKIAA4118
Chromosome Location
Chr. 2: 156065015 - 156069501 on Build GRCm38
UniGene ID
Mm.330713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139151.4 NP_631890.3 10 1135
GenBank mRNA AB041554.1 - 8 1714
AK028080.1 - 10 1104
AK043812.1 - 9 1421
AK220494.1 - 2408

View Full Product Details