Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011933.2 | NP_036063.1 | 8 | 796 | |
GenBank mRNA | AF155575.1 | - | 8 | 796 | |
AK137212.1 | - | 8 | 727 | ||
AK149270.1 | - | 8 | 921 | ||
AK162961.1 | - | 8 | 932 | ||
BC021865.1 | - | 8 | 768 | 5123899 |