Gene Transcripts
- Gene Symbol
- Txnrd2
- Entrez Gene ID
- 26462
- Gene Name
- thioredoxin reductase 2
- Gene Aliases
- AA118373, ESTM573010, TGR, Tr3, Trxr2, Trxrd2
- Chromosome Location
- Chr. 16: 18426417 - 18479073 on Build GRCm38
- UniGene ID
- Mm.390906
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013711.3 |
NP_038739.2
|
5, 6 |
489 |
|
GenBank mRNA |
AB027566.1 |
- |
5, 6 |
489 |
|
AF136399.1 |
- |
5, 6 |
492 |
|
AF171053.1 |
- |
5, 6 |
455 |
|
AF412308.1 |
- |
5, 6 |
491 |
|
AK075828.1 |
- |
5, 6 |
467 |
|
BC013688.1 |
- |
5, 6 |
469 |
3584154 |
BC029344.1 |
- |
5, 6 |
469 |
|
BC052157.1 |
- |
5, 6 |
468 |
6506286 |
BC052758.1 |
- |
5, 6 |
468 |
|
View Full Product Details