Gene Transcripts
- Gene Symbol
- Vps39
- Entrez Gene ID
- 269338
- Gene Name
- vacuolar protein sorting 39 (yeast)
- Gene Aliases
- A230065P22Rik, AW743070, Vam6, Vam6P, mVam6
- Chromosome Location
- Chr. 2: 120316461 - 120353144 on Build GRCm38
- UniGene ID
- Mm.246131
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147153.3 |
NP_671495.1
|
4, 5 |
391 |
|
NM_178851.3 |
NP_849182.1
|
3, 4 |
358 |
|
GenBank mRNA |
AB028844.1 |
- |
3, 4 |
338 |
|
AF281050.1 |
- |
4, 5 |
368 |
|
AF281051.1 |
- |
3, 4 |
335 |
|
AK038824.1 |
- |
3, 4 |
347 |
|
AK042253.1 |
- |
3, 4 |
347 |
|
AK084743.1 |
- |
3, 4 |
358 |
|
AK129210.1 |
- |
3, 4 |
353 |
|
AK140041.1 |
- |
4, 5 |
396 |
|
AK141424.1 |
- |
3, 4 |
341 |
|
AK154103.1 |
- |
3, 4 |
347 |
|
AK159889.1 |
- |
3, 4 |
351 |
|
AK160109.1 |
- |
3, 4 |
351 |
|
AK170831.1 |
- |
4, 5 |
418 |
|
AK171008.1 |
- |
3, 4 |
347 |
|
BB655861.1 |
- |
4, 5 |
384 |
|
BC007479.1 |
- |
3, 4 |
309 |
3492875 |
View Full Product Details