Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012032.4 | NP_036162.3 | 9 | 1282 | |
GenBank mRNA | AB029499.1 | - | 9 | 1191 | |
AF181684.1 | - | 9 | 1210 | ||
AK031101.1 | - | 9 | 1271 | ||
AK128955.1 | - | 7 | 819 | ||
AK145604.1 | - | 8 | 2623 | ||
AK150106.1 | - | 9 | 1228 | ||
AK152720.1 | - | 9 | 1228 | ||
AK155921.1 | - | 9 | 1228 | ||
AK159347.1 | - | 9 | 1228 | ||
AK159488.1 | - | 9 | 1228 | ||
AK159819.1 | - | 9 | 1228 | ||
AK160005.1 | - | 9 | 1228 | ||
AK160055.1 | - | 9 | 1228 | ||
AK161981.1 | - | 9 | 1228 | ||
AK164807.1 | - | 9 | 1228 | ||
AK166924.1 | - | 9 | 1228 | ||
AK170350.1 | - | 9 | 1221 | ||
BC011295.1 | - | 9 | 1238 | 3487943 | |
BC022901.1 | - | 9 | 1219 | 5068196 | |
BC029026.1 | - | 9 | 1247 | 5039323 | |
BC029980.1 | - | 9 | 1251 | ||
L29441.1 | - | 8 | 1007 |