Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017404.4 | NP_059100.3 | 2 | 265 | |
GenBank mRNA | AF239728.1 | - | 2 | 182 | |
AK007681.1 | - | 2 | 201 | ||
AK031942.1 | - | 2 | 186 | ||
BC012274.1 | - | 2 | 144 | 3981658 | |
BC016561.1 | - | 2 | 201 | 2645316 | |
BC036276.1 | - | 2 | 201 |