Gene Transcripts
- Gene Symbol
- Lsm2
- Entrez Gene ID
- 27756
- Gene Name
- LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)
- Gene Aliases
- D17H6S56E-2, D17H6S56E2, Dmapl, Dmpkap, G7b, Sm-X5, SmX5, snRNP
- Chromosome Location
- Chr. 17: 34981854 - 34985891 on Build GRCm38
- UniGene ID
- Mm.165735
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110101.2 |
NP_001103571.1
|
2, 3 |
304 |
|
NM_001204274.1 |
NP_001191203.1
|
2, 3 |
304 |
|
NM_030597.3 |
NP_085100.1
|
2, 3 |
310 |
|
GenBank mRNA |
AF204146.1 |
- |
2, 3 |
281 |
|
AF204147.1 |
- |
2, 3 |
281 |
|
AF204148.1 |
- |
2, 3 |
281 |
|
AF204149.1 |
- |
2, 3 |
281 |
|
AF204150.1 |
- |
2, 3 |
281 |
|
AF204151.1 |
- |
2, 3 |
281 |
|
AF204152.1 |
- |
2, 3 |
281 |
|
AF204153.1 |
- |
1, 2 |
523 |
|
AF204157.1 |
- |
2, 3 |
148 |
|
AK151605.1 |
- |
2, 3 |
278 |
|
AK178855.1 |
- |
2, 3 |
92 |
|
BB848823.1 |
- |
2, 3 |
310 |
|
BC014288.1 |
- |
2, 3 |
289 |
3982410 |
BC049543.1 |
- |
|
303 |
6774564 |
View Full Product Details