Gene Transcripts

Gene Symbol
Kif19a
Entrez Gene ID
286942
Gene Name
kinesin family member 19A
Gene Aliases
Kif19
Chromosome Location
Chr. 11: 114764501 - 114791118 on Build GRCm38
UniGene ID
Mm.308713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102615.1 NP_001096085.1 13, 14 1972
GenBank mRNA AK155544.1 - 13, 14 1936
BC151045.1 - 13, 14 1934 9087958
BC151053.1 - 13, 14 1934 9087966
EF570784.1 - 13, 14 1847

View Full Product Details