Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164491.1 | NP_001157963.1 | 6 | 782 | |
NM_198649.3 | NP_941051.2 | 5 | 1101 | ||
GenBank mRNA | AK086538.1 | - | 5 | 881 | |
AK173040.1 | - | 5 | 659 | ||
BC060275.1 | - | 6 | 753 | 6822626 | |
BC094229.1 | - | 5 | 1075 | 30534236 |