Gene Transcripts

Gene Symbol
Ifitm10
Entrez Gene ID
320802
Gene Name
interferon induced transmembrane protein 10
Gene Aliases
6330512M04Rik, AI848576, DSPA3
Chromosome Location
Chr. 7: 142321583 - 142373781 on Build GRCm38
UniGene ID
Mm.154486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177265.4 NP_796239.1 2 327
GenBank mRNA AK045822.1 - 2 328
AK163573.1 - 2 329
BC049666.1 - 2 317 6534368

View Full Product Details