Gene Transcripts

Gene Symbol
Rab26
Entrez Gene ID
328778
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
A830020M03Rik
Chromosome Location
Chr. 17: 24529054 - 24533747 on Build GRCm38
UniGene ID
Mm.266033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177375.1 NP_796349.1 6, 7 708
GenBank mRNA AB232621.2 - 6, 7 544
AK043912.1 - 5, 6 943
BC094931.1 - 6, 7 616 6436242
BC118018.1 - 6, 7 849 40103722

View Full Product Details