Gene Transcripts

Gene Symbol
Fat4
Entrez Gene ID
329628
Gene Name
FAT tumor suppressor homolog 4 (Drosophila)
Gene Aliases
6030410K14Rik, 9430004M15
Chromosome Location
Chr. 3: 38886487 - 39011987 on Build GRCm38
UniGene ID
Mm.316210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183221.3 NP_899044.3 9 10439
GenBank mRNA BC079887.1 - 1 1533
DQ286572.1 - 9 10439

View Full Product Details