Gene Transcripts

Gene Symbol
Kcp
Entrez Gene ID
333088
Gene Name
kielin/chordin-like protein
Gene Aliases
AW060220, CRIM-2, Crim2, Gm793, KCP-1
Chromosome Location
Chr. 6: 29473162 - 29508011 on Build GRCm38
UniGene ID
Mm.332961
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029985.4 NP_001025156.3 4 447
GenBank mRNA AK154369.1 - 4 435
AY884211.1 - 4 433
BC119794.1 - 4 420
CJ103176.1 - 4 424

View Full Product Details