Gene Transcripts
- Gene Symbol
- Lrrc24
- Entrez Gene ID
- 378937
- Gene Name
- leucine rich repeat containing 24
- Gene Aliases
- 6430402H13Rik
- Chromosome Location
- Chr. 15: 76715276 - 76722173 on Build GRCm38
- UniGene ID
- Mm.289666
- Species
- Mus musculus
- Gene Symbol
- Lrrc14
- Entrez Gene ID
- 223664
- Gene Name
- leucine rich repeat containing 14
- Gene Aliases
- E130306I01Rik, mKIAA0014
- Chromosome Location
- Chr. 15: 76710623 - 76715097 on Build GRCm38
- UniGene ID
- Mm.277582
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042532.1 |
- |
|
421 |
|
AK053747.1 |
- |
|
3132 |
|
View Full Product Details