Gene Transcripts
- Gene Symbol
- Nlrp4d
- Entrez Gene ID
- 384752
- Gene Name
- NLR family, pyrin domain containing 4D
- Gene Aliases
- Nalp-beta, Nalp4d
- Chromosome Location
- Chr. 7: 10358841 - 10388935 on Build GRCm38
- UniGene ID
- Mm.359130
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY596195.1 |
- |
2 |
338 |
|
View Full Product Details