Gene Transcripts

Gene Symbol
Mrgpra5
Entrez Gene ID
404235
Gene Name
MAS-related GPR, member A5
Gene Aliases
MrgA5
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.484774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207534.1 NP_997417.1 456

View Full Product Details