Gene Transcripts

Gene Symbol
Nkain2
Entrez Gene ID
432450
Gene Name
Na+/K+ transporting ATPase interacting 2
Gene Aliases
6330571D19Rik, AW455467, Tcba1
Chromosome Location
Chr. 10: 31689310 - 32893997 on Build GRCm38
UniGene ID
Mm.49665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013411.2 NP_001013429.2 2 370
NM_001025286.2 NP_001020457.2 2 370
GenBank mRNA AY940478.1 - 2 166
BC141063.1 - 2 240 9055675
BY247478.1 - 373
DQ065606.1 - 2 303
DQ065607.1 - 2 303
EF058049.2 - 2 303
EF058050.1 - 1 151
EF058051.1 - 2 151

View Full Product Details