Gene Transcripts
- Gene Symbol
- Psg20
- Entrez Gene ID
- 434540
- Gene Name
- pregnancy-specific glycoprotein 20
- Gene Aliases
- Cea7, EG434540
- Chromosome Location
- Chr. 7: 18674366 - 18685992 on Build GRCm38
- UniGene ID
- Mm.479863
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054058.1 |
NP_473399.1
|
5 |
838 |
|
View Full Product Details