Gene Transcripts

Gene Symbol
Fbxo15
Entrez Gene ID
50764
Gene Name
F-box protein 15
Gene Aliases
AU019763, Fbx15, ecat3
Chromosome Location
Chr. 18: 84934552 - 84981476 on Build GRCm38
UniGene ID
Mm.28369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015798.3 NP_056613.1 8 1111
GenBank mRNA AF176530.1 - 8 1095
AK082809.1 - 8 1132
AK163224.1 - 8 1112
AK166591.1 - 8 1112
AK166599.1 - 8 1113
AK166918.1 - 8 1977
AK167118.1 - 8 1112
AK167270.1 - 8 1111

View Full Product Details