Gene Transcripts
- Gene Symbol
- Mink1
- Entrez Gene ID
- 50932
- Gene Name
- misshapen-like kinase 1 (zebrafish)
- Gene Aliases
- B55, MINK, Map4k6, Ysk2
- Chromosome Location
- Chr. 11: 70562594 - 70614482 on Build GRCm38
- UniGene ID
- Mm.42967
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001045959.1 |
NP_001039424.1
|
11, 12 |
1205 |
|
NM_001045964.1 |
NP_001039429.1
|
11, 12 |
1205 |
|
NM_016713.2 |
NP_057922.2
|
11, 12 |
1205 |
|
NM_176893.2 |
NP_795712.2
|
11, 12 |
1205 |
|
GenBank mRNA |
AB035697.1 |
- |
11, 12 |
1190 |
|
AB041925.1 |
- |
11, 12 |
1191 |
|
BC011346.1 |
- |
6, 7 |
577 |
|
BC052474.1 |
- |
11, 12 |
1060 |
6833893 |
BC079549.1 |
- |
11, 12 |
1063 |
|
CD350084.1 |
- |
4, 5 |
384 |
|
View Full Product Details