Gene Transcripts

Gene Symbol
Wdr72
Entrez Gene ID
546144
Gene Name
WD repeat domain 72
Gene Aliases
AW558070, D230040E23
Chromosome Location
Chr. 9: 74110331 - 74283203 on Build GRCm38
UniGene ID
Mm.335289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033500.3 NP_001028672.2 18 3526
GenBank mRNA AK142559.1 - 18 3506

View Full Product Details