Gene Transcripts

Gene Symbol
Fbxw18
Entrez Gene ID
546161
Gene Name
F-box and WD-40 domain protein 18
Gene Aliases
C85627
Chromosome Location
Chr. 9: 109676732 - 109702700 on Build GRCm38
UniGene ID
Mm.235303
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033794.3 NP_001028966.1 7 970
GenBank mRNA AK136230.1 - 7 974
AK162167.1 - 7 955
BC139081.1 - 7 970 8862103

View Full Product Details