Gene Transcripts

Gene Symbol
Magix
Entrez Gene ID
54634
Gene Name
MAGI family member, X-linked
Gene Aliases
Pdzx, Sfc18
Chromosome Location
Chr. X: 7673166 - 7681251 on Build GRCm38
UniGene ID
Mm.67912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018832.2 NP_061302.2 6 866
GenBank mRNA AF229645.1 - 6 886
AK077898.1 - 6 833
AK158091.1 - 6 806
BC099459.1 - 6 866 30847792
BC132580.1 - 6 805 40130857
BC132582.1 - 6 805 40130859

View Full Product Details