Gene Transcripts

Gene Symbol
Fam60a
Entrez Gene ID
56306
Gene Name
family with sequence similarity 60, member A
Gene Aliases
Ppcs1, Pptcs1, Tera
Chromosome Location
Chr. 6: 148921045 - 148946432 on Build GRCm38
UniGene ID
Mm.18637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019643.3 NP_062617.2 2 242
GenBank mRNA AK046519.1 - 2 149
AK145384.1 - 2 253
AK166569.1 - 2 248
BC076599.1 - 2 242 30614283

View Full Product Details