Gene Transcripts

Gene Symbol
Dnajc4
Entrez Gene ID
57431
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 4
Gene Aliases
2010301J22Rik, Hspf2, Mcg18
Chromosome Location
Chr. 19: 6987911 - 6992696 on Build GRCm38
UniGene ID
Mm.10721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020566.1 NP_065591.1 3 242
GenBank mRNA AF036875.1 - 3 242
AK008507.1 - 3 488
AK170008.1 - 3 481
BC049281.1 - 3 473 4989485

View Full Product Details