Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001029929.2 | NP_001025100.1 | 9 | 2006 | |
GenBank mRNA | AK029988.1 | - | 9 | 2007 | |
AK128979.1 | - | 8 | 2313 | ||
AK171931.1 | - | 9 | 1855 | ||
BC038060.1 | - | 6 | 616 | ||
BC151086.1 | - | 9 | 1678 | 9087999 |