Gene Transcripts
- Gene Symbol
- Trp53inp1
- Entrez Gene ID
- 60599
- Gene Name
- transformation related protein 53 inducible nuclear protein 1
- Gene Aliases
- 2700057G22Rik, SIP, SIP18, SIP27, Stinp, Teap
- Chromosome Location
- Chr. 4: 11156441 - 11174377 on Build GRCm38
- UniGene ID
- Mm.393018
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199105.1 |
NP_001186034.1
|
2, 3 |
355 |
|
NM_021897.3 |
NP_068697.1
|
2, 3 |
355 |
|
GenBank mRNA |
AJ131776.1 |
- |
2, 3 |
356 |
|
AK054214.1 |
- |
2, 3 |
352 |
|
AK142321.1 |
- |
2, 3 |
351 |
|
AY034611.1 |
- |
2, 3 |
356 |
|
AY034612.1 |
- |
2, 3 |
356 |
|
BC132124.1 |
- |
1, 2 |
257 |
40130401 |
BC138711.1 |
- |
1, 2 |
257 |
8861733 |
BC145396.1 |
- |
1, 2 |
256 |
|
View Full Product Details